ENA369382989 Enamine

2-chloro-1-(2,5-difluoro-4-methylphenyl)ethan-1-one

  • MDL number MFCD18910219
  • Cas Number : 1379327-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93...95
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103851

Safety Information

msds https://www.enaminestore.com/catalog/EN300-103851
Availability details for
Quantity
  • Availability
  • Add to Cart