ENA369382992 Enamine

2-(5,6-dimethyl-1H-1,3-benzodiazol-1-yl)propanethioamide

  • MDL number MFCD16077852
  • Cas Number : 1249449-54-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 204...206
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103854

Safety Information

msds https://www.enaminestore.com/catalog/EN300-103854
Availability details for
Quantity
  • Availability
  • Add to Cart