ENA369383003 Enamine

4-amino-N-(oxan-4-yl)benzamide

  • MDL number MFCD12088294
  • Cas Number : 1155632-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 217...219
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103892

Safety Information

msds https://www.enaminestore.com/catalog/EN300-103892
Availability details for
Quantity
  • Availability
  • Add to Cart