ENA369383007 Enamine

(2S)-3-[(3,3,3-trifluoropropyl)amino]propane-1,2-diol

  • MDL number MFCD18298113
  • Cas Number : 1344922-37-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103900

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-103900
Availability details for
Quantity
  • Availability
  • Add to Cart