ENA369383010 Enamine

4-bromo-5-methyl-2,3-dihydro-1H-pyrazol-3-one

  • MDL number MFCD16620112
  • Cas Number : 89363-91-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124...126
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103904

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-103904
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart