ENA369383013 Enamine

4-(aminomethyl)octane

  • MDL number MFCD16781415
  • Cas Number : 857818-18-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103913

Safety Information

msds https://www.enaminestore.com/catalog/EN300-103913
Availability details for
Quantity
  • Availability
  • Add to Cart