ENA369383029 Enamine

4-bromo-6-methyl-2-(propan-2-yl)pyrimidine

  • MDL number MFCD11218845
  • Cas Number : 1134263-20-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104037

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104037
Availability details for
Quantity
  • Availability
  • Add to Cart