ENA369383035 Enamine

5H,6H,7H,8H-[1,2,4]triazolo[1,5-a]pyridin-8-amine

  • MDL number MFCD13176269
  • Cas Number : 132454-36-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109-111
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102652

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102652
Availability details for
Quantity
  • Availability
  • Add to Cart