ENA369383038 Enamine

2-(1-methyl-1H-pyrazol-4-yl)-1,3-oxazole-4-carboxylic acid

  • MDL number MFCD16250906
  • Cas Number : 1247425-58-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102653

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102653
Availability details for
Quantity
  • Availability
  • Add to Cart