ENA369383043 Enamine

1-methoxypentan-3-amine

  • MDL number MFCD08450862
  • Cas Number : 220952-90-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104087

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104087
Availability details for
Quantity
  • Availability
  • Add to Cart