ENA369383047 Enamine

2-(1H-1,3-benzodiazol-2-yl)-2-methylpropan-1-ol

  • MDL number MFCD16859769
  • Cas Number : 1083423-83-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104097

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104097
Availability details for
Quantity
  • Availability
  • Add to Cart