ENA369383049 Enamine

ethyl 4-(3,4-dimethylphenyl)-2,4-dioxobutanoate

  • MDL number MFCD11523438
  • Cas Number : 1019457-35-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104139

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104139
Availability details for
Quantity
  • Availability
  • Add to Cart