ENA369383050 Enamine

3-(5-cyclopropyl-1H-1,2,3,4-tetrazol-1-yl)-4-methoxyaniline

  • MDL number MFCD17298669
  • Cas Number : 1275491-84-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 161...163
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104140

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104140
Availability details for
Quantity
  • Availability
  • Add to Cart