ENA369383055 Enamine

4-(2,5-dichlorothiophen-3-yl)butanoic acid

  • MDL number MFCD11642200
  • Cas Number : 34967-65-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104163

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104163
Availability details for
Quantity
  • Availability
  • Add to Cart