ENA369383056 Enamine

3-(aminomethyl)-1-methylpyrrolidine-2,5-dione hydrochloride

  • MDL number MFCD20542871
  • Cas Number : 86970-11-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102481

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102481
Availability details for
Quantity
  • Availability
  • Add to Cart