ENA369383057 Enamine

2-amino-3-(3,4-difluorophenoxy)-2-methylpropanenitrile

  • MDL number MFCD16135400
  • Cas Number : 1247559-93-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104170

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104170
Availability details for
Quantity
  • Availability
  • Add to Cart