ENA369383059 Enamine

5-phenoxypentanal

  • MDL number MFCD17252877
  • Cas Number : 87841-94-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104177

Safety Information

msds https://www.enaminestore.com/catalog/EN300-104177
Availability details for
Quantity
  • Availability
  • Add to Cart