ENA369383063 Enamine

[2-(1-methoxyethyl)-1,3-thiazol-4-yl]methanamine

  • MDL number MFCD21874400
  • Cas Number : 1376269-88-3
  • Molecular Weight : 172.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-102679

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102679
Availability details for
Quantity
  • Availability
  • Add to Cart