ENA369383074 Enamine

N-(cyclobutylmethyl)aniline

  • MDL number MFCD03211323
  • Cas Number : 191664-09-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102682

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102682
Availability details for
Quantity
  • Availability
  • Add to Cart