ENA369383079 Enamine

1-(2-methyl-1H-imidazol-1-yl)propan-2-ol

  • MDL number MFCD12765013
  • Cas Number : 34091-34-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102684

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-102684
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart