ENA369383089 Enamine

7-amino-6-bromo-3,4-dihydro-2H-1,4-benzoxazin-3-one

  • MDL number MFCD11205104
  • Cas Number : 1094222-68-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 209...211
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102703

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102703
Availability details for
Quantity
  • Availability
  • Add to Cart