ENA369383095 Enamine

1-(2-chloroethyl)-3,5-dimethylcyclohexane

  • MDL number MFCD20405094
  • Cas Number : 20678-16-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102712

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102712
Availability details for
Quantity
  • Availability
  • Add to Cart