ENA369383096 Enamine

N-methyl-N-(2-phenylethyl)pyrrolidin-3-amine

  • MDL number MFCD18325516
  • Cas Number : 1338980-27-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102483

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102483
Availability details for
Quantity
  • Availability
  • Add to Cart