ENA369383101 Enamine

1-(pyridin-3-yl)pentane-1,3-dione

  • MDL number MFCD11185141
  • Cas Number : 622403-56-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104936

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104936
Availability details for
Quantity
  • Availability
  • Add to Cart