ENA369383112 Enamine

4-[(tert-butoxy)methyl]aniline

  • MDL number MFCD11196358
  • Cas Number : 1039311-45-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-102752

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102752
Availability details for
Quantity
  • Availability
  • Add to Cart