ENA369383118 Enamine

1-(3-methyl-2,3-dihydro-1-benzofuran-2-yl)ethan-1-amine hydrochloride

  • MDL number MFCD22196512
  • Cas Number : 1376054-09-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102754

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102754
Availability details for
Quantity
  • Availability
  • Add to Cart