ENA369383123 Enamine

3-methyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carboxylic acid

  • MDL number MFCD06661767
  • Cas Number : 51727-06-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 242-243
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102487

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102487
Availability details for
Quantity
  • Availability
  • Add to Cart