ENA369383125 Enamine

2-(benzylamino)-1-(furan-2-yl)ethan-1-ol

  • MDL number MFCD11937027
  • Cas Number : 146173-92-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68...70
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102764

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102764
Availability details for
Quantity
  • Availability
  • Add to Cart