ENA369383141 Enamine

1-(4-fluorophenyl)-3-hydroxypropan-2-one

  • MDL number MFCD16165842
  • Cas Number : 565428-06-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102488

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102488
Availability details for
Quantity
  • Availability
  • Add to Cart