ENA369383167 Enamine

{[1-(3,5-dimethoxyphenyl)-1H-1,2,3,4-tetrazol-5-yl]methyl}(methyl)amine hydrochloride

  • MDL number MFCD22196488
  • Cas Number : 1376395-33-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 231...233
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102489

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102489
Availability details for
Quantity
  • Availability
  • Add to Cart