ENA369383170 Enamine

4-amino-2-chlorobenzene-1-thiol

  • MDL number MFCD11635112
  • Cas Number : 15178-53-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40437

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40437
Availability details for
Quantity
  • Availability
  • Add to Cart