ENA369383174 Enamine

1-[3-(diethylamino)propyl]-2,5-dihydro-1H-pyrrole-2,5-dione

  • MDL number MFCD11149219
  • Cas Number : 5135-67-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44381

Safety Information

msds https://www.enaminestore.com/catalog/EN300-44381
Availability details for
Quantity
  • Availability
  • Add to Cart