ENA369383218 Enamine

1-(5-ethyl-1,2,4-oxadiazol-3-yl)cyclopentan-1-amine

  • MDL number MFCD12138416
  • Cas Number : 1153835-34-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-63748

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63748
Availability details for
Quantity
  • Availability
  • Add to Cart