ENA369383223 Enamine

1,1,1-trifluoro-3-nitropropan-2-ol

  • MDL number MFCD01328287
  • Cas Number : 453-35-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-64210

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64210
Availability details for
Quantity
  • Availability
  • Add to Cart