ENA369383227 Enamine

2-amino-6-chloro-3-fluorobenzoic acid

  • MDL number MFCD11183542
  • Cas Number : 1039815-76-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 161...163
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65909

Safety Information

msds https://www.enaminestore.com/catalog/EN300-65909
Availability details for
Quantity
  • Availability
  • Add to Cart