ENA369383231 Enamine

5-bromo-4-methoxy-2-(piperazin-1-yl)pyrimidine

  • MDL number MFCD07781226
  • Cas Number : 885267-38-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89...91
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68539

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68539
Availability details for
Quantity
  • Availability
  • Add to Cart