ENA369383234 Enamine

2-(4-methanesulfonylphenyl)propanoic acid

  • MDL number MFCD14602272
  • Cas Number : 1250933-90-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69255

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69255
Availability details for
Quantity
  • Availability
  • Add to Cart