ENA369383254 Enamine

4-methanesulfinylbenzaldehyde

  • MDL number MFCD18253462
  • Cas Number : 37794-15-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75887

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75887
Availability details for
Quantity
  • Availability
  • Add to Cart