ENA369383278 Enamine

1-methylcyclobutan-1-ol

  • MDL number MFCD01218890
  • Cas Number : 20117-47-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83389

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-83389
pictograms GHS02,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart