ENA369383279 Enamine

tert-butyl N-(1-oxo-2,3-dihydro-1H-inden-5-yl)carbamate

  • MDL number MFCD22056444
  • Cas Number : 1116358-96-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175-176
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83455

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83455
Availability details for
Quantity
  • Availability
  • Add to Cart