ENA369383286 Enamine

ethyl 5-amino-3-(methylsulfanyl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD01108198
  • Cas Number : 59541-46-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132...134
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84758

Safety Information

msds https://www.enaminestore.com/catalog/EN300-84758
Availability details for
Quantity
  • Availability
  • Add to Cart