ENA369383288 Enamine

6,8-dichloro-1,2,3,4-tetrahydroisoquinoline hydrochloride

  • MDL number MFCD13181513
  • Cas Number : 73075-50-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 292...294
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102833

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-102833
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart