ENA369383310 Enamine

1,3-benzothiazole-6-sulfonyl chloride

  • MDL number MFCD03086100
  • Cas Number : 181124-40-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116...118
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88652

Safety Information

msds https://www.enaminestore.com/catalog/EN300-88652
Availability details for
Quantity
  • Availability
  • Add to Cart