ENA369383315 Enamine

3-(2-aminophenyl)-5-(trifluoromethyl)-1,3-oxazolidin-2-one

  • MDL number MFCD22196667
  • Cas Number : 1376282-88-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90892

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90892
Availability details for
Quantity
  • Availability
  • Add to Cart