ENA369383321 Enamine

5-chloro-2-sulfanylbenzonitrile

  • MDL number MFCD12756632
  • Cas Number : 38244-31-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102839

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102839
Availability details for
Quantity
  • Availability
  • Add to Cart