ENA369383322 Enamine

4-[(5-ethylfuran-2-yl)carbonyl]aniline

  • MDL number MFCD20325588
  • Cas Number : 1343032-79-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124...126
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102846

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102846
Availability details for
Quantity
  • Availability
  • Add to Cart