ENA369383356 Enamine

2-[(4-chlorophenyl)carbonyl]pyrazine

  • MDL number MFCD00160526
  • Cas Number : 118543-85-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102852

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102852
Availability details for
Quantity
  • Availability
  • Add to Cart