ENA369383363 Enamine

bis(3-methylphenyl)methanamine

  • MDL number MFCD12154069
  • Cas Number : 858453-29-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102856

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102856
Availability details for
Quantity
  • Availability
  • Add to Cart