ENA369383376 Enamine

4-chloro-2-[3-(propan-2-yl)-1,2,4-oxadiazol-5-yl]aniline

  • MDL number MFCD11124605
  • Cas Number : 1040079-81-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102494

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102494
Availability details for
Quantity
  • Availability
  • Add to Cart