ENA369383377 Enamine

N-(2-aminoethyl)-1-(1,2-oxazol-3-yl)methanesulfonamide

  • MDL number MFCD22056458
  • Cas Number : 1376280-75-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94969

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-94969
Availability details for
Quantity
  • Availability
  • Add to Cart